Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1, p Thr292 Antibody (SR1773), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | MEK1 |
---|---|
Clone | SR1773 |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
MEK1 Monoclonal antibody specifically detects MEK1 in Human samples. It is validated for Western BlotSpecifications
MEK1 | |
Western Blot 1:500-1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
A synthesized peptide derived from human phospho-MEK1 [Thr292] (Uniprot #: Q02750 ) | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
SR1773 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5604 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title