Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572416
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2416 10 μL
50-157-2415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2416 Supplier Bethyl Laboratories, Inc Supplier No. IHC00619T
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Pig, Panda, Monkey, Gorill

MAP2K1 (MEK1) is a dual specificity mitogen-activated protein kinase (MAPK) kinase. It is an essential component of the MAP kinase pathway and stimulates the enzymatic activity of MAP kinases in response to various signals. Growth factors, cytokines, and proto-oncogenes transduce their signals through the activation of RAS, which leads to activation of the serine/threonine kinase RAF, and then to activation of MAPK kinase. MAP2K1 is activated by the Raf family member p74raf-1, through phosphorylation of serine residues at positions 217 and 221 in the activation loop.

Specifications

Antigen MEK1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAP2K1
Gene Accession No. Q02750
Gene Alias MAPKK1, MEK1, MKK1, PRKMK1
Gene Symbols MAP2K1
Host Species Rabbit
Immunogen Between 343 and 393
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.