Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEK1 Recombinant Rabbit Monoclonal Antibody (BLR095G)
GREENER_CHOICE

Catalog No. PIMA544381
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544381 100 μL
PIMA544382 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544381 Supplier Invitrogen™ Supplier No. MA544381
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

MEK1 Recombinant Monoclonal Antibody for Flow, IP, Western Blot

MAP2K1 (MEK1) kinase is a dual-specificity protein kinase that functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation. MAP2K1 is activated by a wide variety of growth factors and cytokines and also by membrane-dependent depolarization and calcium influx. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BLR095G
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6
Gene Map2k1
Gene Accession No. P31938, Q02750
Gene Alias CFC3; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; Map2k1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg
Gene Symbols Map2k1
Host Species Rabbit
Immunogen Peptide representing a region between residues 1 and 50 of human dual specificity mitogen-activated protein kinase kinase 1 (MEK1).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26395, 5604
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.