Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK2 Antibody (OTI1A2), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272647FR
Description
MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
MEK2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
MAP2K2 | |
Full length recombinant protein of human MEK2 (NP_109587) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2a |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2 | |
Mouse | |
Affinity Purified | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction | |
5605.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction