Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Mouse anti-Human, Clone: 4C3, Bioss
SDP

Catalog No. 501979939
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9939 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9939 Supplier Bioss Supplier No. BSM33216M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEK1 (Mitogen activated protein kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.

Specifications

Antigen MEK2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C3
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MAP2K2
Gene Accession No. P36507
Gene Alias AA589381; CFC4; dual specificity mitogen activated protein kinase kinase 2; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; ERK kinase 2; FLJ26075; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP2K1; Map2k2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK2; MEK 1; MEK 2; MEK-1; Mek2; MEK-2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; MK2; MKK1; Mkk2; OTTHUMP00000165827; PRKMK1; Prkmk2; protein kinase, mitogen activated, kinase 2, p45
Gene Symbols MAP2K2
Host Species Mouse
Immunogen Recombinant human MEK2 Protein.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5605
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.