Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK2 Rabbit anti-Human, Mouse, Clone: 6D5K1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | MEK2 |
---|---|
Clone | 6D5K1 |
Applications | Western Blot, KnockDown |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
MEK2 Monoclonal antibody specifically detects MEK2 in Human, Mouse samples. It is validated for Western Blot, KnockDownSpecifications
MEK2 | |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Kinase, Protein Phosphatase, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
5605 | |
IgG | |
Affinity purified |
6D5K1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEK2 (P36507). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title