Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK5 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP197478AF750
Description
MEK5 Polyclonal specifically detects MEK5 in Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MEK5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
HsT17454, MAP kinase kinase 5, MAP kinase kinase MEK5b, MAPK/ERK kinase 5, MAPKK 5, MAPKK5EC 2.7.12.2, MEK5MEK 5, mitogen-activated protein kinase kinase 5, MKK5, PRKMK5dual specificity mitogen-activated protein kinase kinase 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP2K5 | |
Synthetic peptide corresponding to a portion of human MEK5. | |
0.1 mL | |
Neuroscience, Phospho Specific, Protein Kinase | |
Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction