Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP19747801 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19747801 0.1 mg
NB018780 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP19747801 Supplier Novus Biologicals Supplier No. NBP1974780.1MG
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEK5 Polyclonal specifically detects MEK5 in Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MEK5
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Alias HsT17454, MAP kinase kinase 5, MAP kinase kinase MEK5b, MAPK/ERK kinase 5, MAPKK 5, MAPKK5EC 2.7.12.2, MEK5MEK 5, mitogen-activated protein kinase kinase 5, MKK5, PRKMK5dual specificity mitogen-activated protein kinase kinase 5
Gene Symbols MAP2K5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a portion of human MEK5.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5607
Target Species Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus species
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.