Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MEK5 Recombinant Rabbit Monoclonal Antibody (7B7O10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535172
Description
Immunogen sequence: DVYRKMPEHV LGRIAVAVVK GLTYLWSLKI LHRDVKPSNM LVNTRGQVKL CDFGVSTQLV NSIAKTYVGT NAYMAPERIS GEQYGIHSDV WSLGISFMEL A.
MAP2K5 (MEK5) serine/threonine kinase (mitogen-activated protein kinase 5) or MEK5 is a dual specificity protein kinase that belongs to the MAP kinase family which specifically interacts with and activates MAPK7/ERK5. MAP2K5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation. The rs3743354 polymorphism in the MAP2K5 promoter may affect the risk of developing colorectal cancer (CRC).
Specifications
MEK5 | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13163, Q62862, Q9WVS7 | |
MAP2K5 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human MEK5 (Q13163). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
7B7O10 | |
Unconjugated | |
MAP2K5 | |
AI324775; AI428457; Dual specificity mitogen-activated protein kinase kinase 5; HsT17454; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAP2K5; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; Mapkk5; MEK 5; MEK5; mitogen activated protein kinase kinase 5; mitogen-activated protein kinase kinase 5; Mkk5; PRKMK5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23938, 29568, 5607 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction