Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEK5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529236 Supplier Invitrogen™ Supplier No. PA529236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (98%), Rat (98%), Chicken (91%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MAP2K5 (MEK5) serine/threonine kinase (mitogen-activated protein kinase 5) or MEK5 is a dual specificity protein kinase that belongs to the MAP kinase family which specifically interacts with and activates MAPK7/ERK5. MAP2K5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation. The rs3743354 polymorphism in the MAP2K5 promoter may affect the risk of developing colorectal cancer (CRC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MAP2K5
Gene Accession No. Q13163
Gene Alias AI324775; AI428457; Dual specificity mitogen-activated protein kinase kinase 5; HsT17454; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAP2K5; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; Mapkk5; MEK 5; MEK5; mitogen activated protein kinase kinase 5; mitogen-activated protein kinase kinase 5; Mkk5; PRKMK5
Gene Symbols MAP2K5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MEK5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5607
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.