Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mekk-1 Polyclonal Antibody, Abbomax

Catalog No. 502203816
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3816 Supplier Abbomax Inc Supplier No. 5007884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human uterine carcinoma, thyroid carcinoma Cellular location: Cytoplasmic.

Mekk-1 Polyclonal antibody specifically detects Mekk-1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Mekk-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene MAP3K1
Gene Accession No. P53349, Q13233, Q62925
Gene Alias MAP/ERK kinase kinase 1; MAP3K1; MAPK/ERK kinase kinase 1; MAPKKK1; MEK kinase 1; MEKK; MEKK 1; Mekk1; mitogen activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase; si:rp71-80a19.2; SRXY6
Host Species Rabbit
Immunogen A synthetic peptide derived from internal segment of human Mekk-1
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116667, 26401, 4214
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.