Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111232 Supplier Invitrogen™ Supplier No. PA5111232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSSKCDDSFG CSSNSSNAVI PSDETVFTPV EEKCRLDVNT ELNSSIEDLL EASMPSSDTT VTFKSEVAVL SPEKAENDDT YKDDVNHNQK CKEKMEAEEE EALAIAMAMS ASQDALPIVP QLQVENGEDI IIIQQ Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 91%.

Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK1 can phosphorylate and activate MAPKK 1 and MAPKK 2 (MEK1/MEK2) which leads to phosphorylation of MAP kinases. It is also a highly efficient activator of the JNK cascade. The protein contains a putative 1 RING-type zinc finger and 1 SWIM-type zinc finger.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MAP3K1
Gene Accession No. Q13233
Gene Alias MAP/ERK kinase kinase 1; MAP3K1; MAPK/ERK kinase kinase 1; MAPKKK1; MEK kinase 1; MEKK; MEKK 1; Mekk1; mitogen activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase; si:rp71-80a19.2; SRXY6
Gene Symbols MAP3K1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MEKK1. Recombinant protein control fragment (Product #RP-106119).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4214
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.