Learn More
Invitrogen™ MEKK2 Monoclonal Antibody (4B4)

Description
MA5-15908 targets MAP3K2 in indirect ELISA, ICC, IF and WB applications and shows reactivity with Human samples. The MA5-15908 immunogen is purified recombinant fragment of human MAP3K2 expressed in E. Coli. . MA5-15908 detects MAP3K2 which has a predicted molecular weight of approximately 70kDa.
Specifications
Specifications
| Antigen | MEKK2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 4B4 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | Map3k2 |
| Gene Accession No. | Q61083, Q9Y2U5 |
| Gene Alias | 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.