Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554443
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554443 Supplier Invitrogen™ Supplier No. PA554443
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAFGQSMDLH YTNNELVIPL TTQDDLDKAV ELLDRSIHMK SLKILLVING STQATNLEPL PSLEDLDNTV FGAERKKRLS IIGPTSRDRS SPPPGYIPDE LHQVARNGSF TSINSEGEFI PESMDQM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Map3k2
Gene Accession No. Q9Y2U5
Gene Alias 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b
Gene Symbols Map3k2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEKK2. Recombinant protein control fragment (Product #RP-102059).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.