Learn More
Invitrogen™ MEKK2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590246
Description
Immunogen sequence: MDDQQALNSI MQDLAVLHKA SRPALSLQET RKAKSSSPKK QNDVRVKFEH RGEKRILQFP RPVKLEDLRS KAKIAFGQSM DLHYTNNELV IPLTTQDDLD KAVELLDRSI HMKSLKILLV INGSTQATNL EPLPSLEDLD NTVFGAERKK RLSIIGPTSR DRSSPPPGYI PDELHQVARN; Positive Samples: mouse liver; Cellular Location: Cytoplasm, Nucleus.
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
Specifications
MEKK2 | |
Polyclonal | |
Unconjugated | |
Map3k2 | |
2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26405 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q61083 | |
Map3k2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human MEKK2 (NP_0066003). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.