Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595003
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595003 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595003 Supplier Invitrogen™ Supplier No. PA595003
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Caco-2 whole cell, human K562 whole cell, human THP-1 whole cell, human Raji whole cell, rat kidney tissue, rat brain tissue, rat heart tissue, rat C6 whole cell, mouse kidney tissue, mouse brain tissue, mouse heart tissue, mouse Neuro-2a whole cell, mouse NIH/3T3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK2
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Map3k2
Gene Accession No. Q61083, Q9Y2U5
Gene Alias 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b
Gene Symbols Map3k2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human MEKK2 (606-619aa SADELLRHMFVHYH).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10746, 171492, 26405
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.