Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEKK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | MEKK2 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5986
![]() ![]() |
Proteintech Group Inc
551061AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The antibody is specific to MAP3K2.
The protein encoded by this gene is a member of serine/threonine protein kinase family. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.Specifications
MEKK2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q61083, Q9Y2U5 | |
10746, 171492, 26405 | |
Peptide | |
Primary | |
-20°C | |
MAP3K2 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MAP3K2, MAPK/ERK kinase kinase 2, MAPKKK2, MEK kinase 2, MEKK 2, MEKK2, MEKK2B | |
Map3k2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title