Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEKK3 Antibody (2H11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MEKK3 |
---|---|
Clone | 2H11 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
MEKK3 Monoclonal antibody specifically detects MEKK3 in Human samples. It is validated for ELISASpecifications
MEKK3 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11.25, MAP/ERK kinase kinase 3, MAPK/ERK kinase kinase 3, MAPKKK3MEKK 3, MEK kinase 3, MEKK3, mitogen-activated protein kinase kinase kinase 3 | |
A synthesized peptide derived from Human MEKK3 [UniProt Q99759] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
2H11 | |
ELISA | |
Unconjugated | |
Rabbit | |
Angiogenesis, Cancer, Immunology, Innate Immunity, Protein Kinase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4215 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title