Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK3 Recombinant Rabbit Monoclonal Antibody (9N3Y9)
GREENER_CHOICE

Catalog No. PIMA538090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538090 Supplier Invitrogen™ Supplier No. MA538090
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, NIH/3T3, Jurkat, SH-SY5Y, Mouse spleen Immunogen sequence: MDEQEALNSI MNDLVALQMN RRHRMPGYET MKNKDTGHSN RQSDVRIKFE HNGERRIIAF SRPVKYEDVE HKVTTVFGQP LDLHYMNNEL SILLKNQDDL.

MAP3K3 (MEKK3) directly regulates MAPK8 and extracellular signal-regulated protein kinase (ERK) pathways by phosphorylating multiple MAP2Ks. Northern blot analysis revealed a 4. 6-kb transcript that appears to be ubiquitously expressed. This protein directly regulates the stress-activated protein kinase (SAPK) and extracellular signal-regulated protein kinase (ERK) pathways by activating SEK and MEK1/2 respectively; it does not regulate the p38 pathway. In cotransfection assays, it enhanced transcription from a nuclear factor kappa-B (NFKB)-dependent reporter gene, consistent with a role in the SAPK pathway. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9N3Y9
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Map3k3
Gene Accession No. Q61084, Q99759
Gene Alias AW548911; MAP/ERK kinase kinase 3; MAP3K3; MAPK/ERK kinase kinase 3; MAPKKK3; MEK kinase 3; MEKK 3; Mekk3; mitogen activated protein kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase 3; mKIAA4031
Gene Symbols Map3k3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEKK3 (Q99759).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26406, 4215
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.