Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEKK4 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232187B
Description
MEKK4 Polyclonal antibody specifically detects MEKK4 in Human samples. It is validated for Western BlotSpecifications
MEKK4 | |
Polyclonal | |
Western Blot | |
EC 2.7.11.25, FLJ42439, MAPKKK4yeast, homolog of, MEKK 4, mitogen-activated protein kinase kinase kinase 4 | |
The immunogen this antibody was made to, maps to a region between residue 75 to 125 of human Mitogen-Activated Protein Kinase Kinase Kinase 4 using the numbering given in entry NP_005913.2 (GeneID 4216). | |
0.1 mL | |
Angiogenesis, Cancer, Phospho Specific, Protein Kinase | |
4216 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Biotin | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction