Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEKK6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515093
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515093 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515093 Supplier Invitrogen™ Supplier No. PA515093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEKK6 Polyclonal Antibody for Western Blot, IHC (P)

Mitogen-activated protein kinase (MAPK) signaling cascades include MAPK or extracellular signal-regulated kinase (ERK), MAPK kinase (MKK or MEK), and MAPK kinase kinase (MAPKKK or MEKK). MAPKK kinase/MEKK phosphorylates and activates its downstream protein kinase, MAPK kinase/MEK, which in turn activates MAPK. The kinases of these signaling cascades are highly conserved, and homologs exist in yeast, Drosophila, and mammalian cells. MEKK6 activates the JNK, but not ERK or p38 kinase pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEKK6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MAP3K6
Gene Accession No. O95382, Q9WTR2
Gene Alias apoptosis signal-regulating kinase 2; ASK2; map3k6; MAPKKK6; MEKK6; MGC125653; MGC20114; mitogen activated protein kinase kinase kinase 6; mitogen-activated protein kinase kinase kinase 6; OTTHUMP00000005063
Gene Symbols MAP3K6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1028-1058 amino acids from the C-terminal region of human MEKK6.
Purification Method Ammonium Sulfate Precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53608, 9064
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.