Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEL-18 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204032AF750
Description
MEL-18 Polyclonal antibody specifically detects MEL-18 in Human samples. It is validated for Western BlotSpecifications
MEL-18 | |
Polyclonal | |
Western Blot | |
MEL-18, polycomb group ring finger 2, polycomb group RING finger protein 2, RING finger protein 110DNA-binding protein Mel-18, RNF110MEL18, Zinc finger protein 144, ZNF144MGC10545 | |
The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human DNA-binding Protein Mel-18 using the numbering given in entry NP_009075.1 (GeneID 7703). | |
0.1 mL | |
Zinc Finger | |
7703 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction