Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MEL18 Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA520275
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA520275 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA520275 Supplier Invitrogen™ Supplier No. MA520275
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LTLATVPTPS EGTNTSGASE CESVSDKAPS PATLPATSSS LPSPATPSHG SPSSHGPPA.

The protein encoded by this gene contains a RING finger motif and is similar to the polycomb group (PcG) gene products. PcG gene products form complexes via protein-protein interaction and maintain the transcription repression of genes involved in embryogenesis, cell cycles, and tumorigenesis. This protein was shown to act as a negative regulator of transcription and has tumor suppressor activity. The expression of this gene was detected in various tumor cells, but is limited in neural organs in normal tissues. Knockout studies in mice suggested that this protein may negatively regulate the expression of different cytokines, chemokines, and chemokine receptors, and thus plays an important role in lymphocyte differentiation and migration, as well as in immune responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEL18
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E4
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PCGF2
Gene Accession No. P35227
Gene Alias DNA-binding protein Mel-18; Mel18; Mel-18; Melanoma nuclear protein 18; MGC10545; Pcgf2; polycomb group ring finger 2; polycomb group RING finger protein 2; RING finger protein 110; RNF110; Zfp144; Zfp-144; Zinc finger protein 144; zinc finger protein 144 (Mel-18) (human homolog); ZNF144
Gene Symbols PCGF2
Host Species Mouse
Immunogen PCGF2 (AAH04858.1, 236 a.a. ~ 294 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7703
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.