Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanesulfonyl chloride, 99.5%
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -33.0°C |
Density | 1.4800g/mL |
Boiling Point | 161.0°C |
Flash Point | 110°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.4% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.4435 to 1.4455 |
Linear Formula | CH3SO2Cl |
Quantity | 250 mL |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Toxic in contact with skin. May cause an allergic skin reaction. Fatal if inhaled. Causes severe skin burns and eye damage. Toxic if swallowed. May cause respiratory irritation. May be corrosive |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at |
EINECSNumber | 204-706-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction