Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melamine Mouse anti-Human, Clone: 1B12, Bioss
SDP

Catalog No. 501979913
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9913 Supplier Bioss Supplier No. BSM2182M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melamine is combined with formaldehyde to produce melamine resin, a very durable thermosetting plastic used in Formica, and melamine foam, a polymeric cleaning product. The end products include countertops, dry erase boards, fabrics, glues, housewares and flame retardants. Melamine is one of the major components in Pigment Yellow 150, a colorant in inks and plastics.Melamine also enters the fabrication of melamine poly-sulfonate used as superplasticizer for making high-resistance concrete. Sulfonated melamine formaldehyde (SMF) is a polymer used as cement admixture to reduce the water content in concrete while increasing the fluidity and the workability of the mix during its handling and pouring. It results in concrete with a lower porosity and a higher mechanical strength exhibiting an improved resistance to aggressive environments and a longer life-time. The use of melamine as fertilizer for crops had been envisaged during the ′50s and ′60s because of its high nitrogen content (2/3).[7] However melamine is much more expensive to produce than other common nitrogen fertilizers, such as urea. To be effective as a fertilizer, it is essential that the plant nutrients are released or made available in a manner that matches the needs of the growing crop. The nitrogen mineralization process for melamine is extremely slow, making this product both economically and scientifically impractical for use as a fertilizer.

Specifications

Antigen Melamine
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1B12
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Mouse
Immunogen OVA conjugated Melamine modified.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.