Learn More
Invitrogen™ Melan-A Monoclonal Antibody (CL12864)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA562997
Description
Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 67%.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
| Melan-A | |
| Monoclonal | |
| 0.97 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16655 | |
| MLANA | |
| Recombinant protein corresponding to Human MLANA. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CL12864 | |
| Unconjugated | |
| MLANA | |
| A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
| Mouse | |
| Protein A | |
| RUO | |
| 2315 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.