Learn More
Invitrogen™ Melan-A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120982
Description
Positive test controls include: M21, Mouse eye, Rat eye. The target is usually found in the following locations: Endoplasmic reticulum membrane, Golgi apparatus, Melanosome, Single-pass type III membrane protein, trans-Golgi network membrane. Immunogen sequence: CRRRNGYRAL MDKSLHVGTQ CALTRRCPQE GFDHRDSKVS LQEKNCEPVV PNAPPAYEKL SAEQSPPPYS P.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
Melan-A | |
Polyclonal | |
Unconjugated | |
MLANA | |
A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2315, 293890, 77836 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1.08 mg/mL | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q16655 | |
MLANA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 48-118 of human Melan-A/MART-1 (NP_0055021). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.