Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Melan-A Recombinant Rabbit Monoclonal Antibody (8U1D0)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551353
Description
Immunogen sequence: VLLLIGCWYC RRRNGYRALM DKSLHVGTQC ALTRRCPQEG FDHRDSKVSL QEKNCEPVVP NAPPAYEKLS AEQSPPPYSP.
MART1 is involved in melanosome biogenesis by ensuring the stability of GPR143. Plays a vital role in the expression, stability, trafficking, and processing of melanocyte protein PMEL, which is critical to the formation of stage II melanosomes.
Specifications
Melan-A | |
Recombinant Monoclonal | |
0.08 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q16655 | |
MLANA | |
A synthetic peptide corresponding to a sequence within amino acids 11-110 of human MLANA (NP_005502.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
8U1D0 | |
Unconjugated | |
MLANA | |
A930034P04Rik; Antigen LB39-AA; antigen SK29-AA; MART1; MART-1; melan-A; melanoma antigen recognized by T-cells 1; melanoma associated antigen recognized by T-cells 1; MLANA; Protein Melan-A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2315 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction