Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-1 R/MC1R Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30385820UL
Description
Melanocortin-1 R/MC1R Polyclonal specifically detects Melanocortin-1 R/MC1R in Human, Mouse samples. It is validated for Western Blot.Specifications
Melanocortin-1 R/MC1R | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
Unconjugated | |
CMM5, MC1-R, melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor), Melanocortin receptor 1, melanocyte-stimulating hormone receptor, melanotropin receptor, MSHR, MSH-RMGC14337, SHEP2 | |
Recombinant protein of human Melanocortin-1 R/MC1R. | |
20 μL | |
GPCR, Neuroscience | |
4157 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction