Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-3 R/MC3R Antibody (345404), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3737W
Description
Melanocortin-3 R/MC3R Monoclonal antibody specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
Melanocortin-3 R/MC3R | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
345404 | |
Flow Cytometry, CyTOF-ready | |
BMIQ9, MC3, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, OB20, Obesity quantitative trait locus, OQTL | |
Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R | |
0.1 mL | |
GPCR | |
4159 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction