Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-3 R/MC3R Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001501
Description
Melanocortin-3 R/MC3R Polyclonal specifically detects Melanocortin-3 R/MC3R in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
Melanocortin-3 R/MC3R | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BMIQ9, MC3, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, OB20, Obesity quantitative trait locus, OQTL | |
Goat | |
Affinity Purified | |
RUO | |
4159 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_063941. | |
MC3R | |
Peptide with sequence SIQKTYLEGDFV-C corresponding to N-Terminus according to NP_063941.2. | |
0.1 mg | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction