Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-4 R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241103
Description
Melanocortin-4 R Polyclonal specifically detects Melanocortin-4 R in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Melanocortin-4 R | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
MC4R | |
Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human MC4R. The immunogen is located within the first 50 amino acids of MC4R. Amino Acid Squence: SYRLHSNASESLGKGYSDG | |
0.1 mg | |
GPCR, Neuroscience | |
4160 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 ug/ml, ELISA, Immunohistochemistry 2.5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 2.5 ug/ml | |
MC4-R, melanocortin 4 receptor, melanocortin receptor 4, MGC126851, MGC138197 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction