Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-4 R Antibody (SR1620), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321874100UL
Description
Melanocortin-4 R Monoclonal antibody specifically detects Melanocortin-4 R in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Melanocortin-4 R | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1620 | |
Western Blot 1:500-1:2000 | |
MC4-R, melanocortin 4 receptor, melanocortin receptor 4, MGC126851, MGC138197 | |
A synthesized peptide derived from human Melanocortin-4 R (Uniprot #: P32245) | |
100 μg | |
GPCR, Neuroscience | |
4160 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction