Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-5 R/MC5R Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NLS1024
Description
Melanocortin-5 R/MC5R Polyclonal specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Melanocortin-5 R/MC5R | |
Polyclonal | |
Unconjugated | |
PBS with 0.1% Sodium Azide | |
MC5R | |
Synthetic 16 amino acid peptide from 1st extracellular domain of human Melanocortin-5 R/MC5R. | |
0.05 mL | |
GPCR | |
4161 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry, Immunohistochemistry-Paraffin 20 ug/ml | |
MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Melanocortin 5 Receptor | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction