Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanocortin-5 R/MC5R Antibody (SR1772), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32248150UL
Description
Melanocortin-5 R/MC5R Monoclonal antibody specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Melanocortin-5 R/MC5R | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1772 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor | |
A synthesized peptide derived from human Melanocortin-5 R/MC5R (Uniprot #: P33032) | |
50 μg | |
GPCR | |
4161 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction