Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanoma Associated Antigen Antibody (MAA/1414), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308848AF350
Description
Melanoma Associated Antigen Monoclonal antibody specifically detects Melanoma Associated Antigen in Human samples. It is validated for ELISASpecifications
Melanoma Associated Antigen | |
Monoclonal | |
Alexa Fluor 350 | |
Chronic μgcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like | |
proteins extracted from a melanoma | |
100 μg | |
Primary | |
Store at 4°C in the dark. | |
IgG2b κ |
ELISA | |
MAA/1414 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction