Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanoma Associated Antigen Mouse anti-Human, Clone: MAA/1414, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Melanoma Associated Antigen |
---|---|
Clone | MAA/1414 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 1-5 ug/ml |
Applications | ELISA |
Description
Melanoma Associated Antigen Monoclonal specifically detects Melanoma Associated Antigen in Human samples. It is validated for ELISA.Specifications
Melanoma Associated Antigen | |
0.2 mg/ml | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
proteins extracted from a melanoma | |
Primary | |
Store at 4°C. |
MAA/1414 | |
ELISA 1-5 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title