Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanoma Associated Antigen (PNL2) Antibody (PNL2), FITC , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311561F
Description
Melanoma Associated Antigen (PNL2) Monoclonal antibody specifically detects Melanoma Associated Antigen (PNL2) in Human, Mouse, Canine samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Melanoma Associated Antigen (PNL2) | |
Monoclonal | |
FITC | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse, Canine | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PNL2 | |
PBS | |
Melanocyte antigen | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction