Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Melanoma Discriminated Antigen Monoclonal Antibody (PAL-M1)
GREENER_CHOICE

Catalog No. 01671052
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-052 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-052 Supplier Invitrogen™ Supplier No. MA135614
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody PAL-M1 stains the majority of primary melanomas and the large majority of metastases and is directed against the transferrin receptor (CD71). Local staining of dysplastic nevocellular nevi may be observed. Common nevocellular nevi rarely stain.

Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths. Despite many years of intensive laboratory and clinical research, the sole effective cure is surgical resection of the primary tumor before it achieves a thickness of greater than 1 mm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melanoma Discriminated Antigen
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone PAL-M1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 10mM sodium azide
Gene Alias melanoma antigen; melanoma associated antigen
Host Species Mouse
Immunogen Human melanoma metastases tissue.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.