Learn More
Invitrogen™ Melanoma Discriminated Antigen Monoclonal Antibody (PAL-M2)

Description
This antibody stains a minority of primary melanomas and half of the metastatic lesions tested. It rarely stains dysplastic naevi or common cellular naevi using standard immunohistochemical conditions. The antibody recognizes two protein bands in immunoblotting with a molecular weight of 95-100 kD.
Specifications
Specifications
| Antigen | Melanoma Discriminated Antigen |
| Applications | Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | PAL-M2 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 10mM sodium azide |
| Gene Alias | melanoma antigen; melanoma associated antigen |
| Host Species | Mouse |
| Immunogen | Isolated from Human Melanoma cells. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.