Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS364S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS364S0 100 μL Concentrated
MS364S1 500 μL Concentrated
MS364S 1 mL Concentrated
MS364R7 7 mL Prediluted
MS364RQ 12 mL Ready-to-Use
MS364R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS364S0 Supplier Epredia™ Supplier No. MS364S0
Add to Cart
Add to Cart

Employ one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available wit the Epredia™ Melanoma (gp100) Ab-1, Mouse Monoclonal Antibody.

Research Areas: Cancer Marker

Host Species: Mouse

Clone: HMB45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with dog and rat. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes

Molecular Weight: 100kDa

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

    Western Blotting

Specifications

Antigen Melanoma (gp100) Ab-1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone HMB45
Conjugate Unconjugated
Host Species Mouse
Immunogen Extract of pigmented melanoma metastases from lymph nodes
Quantity 100 μL
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.