Learn More
Description
Host Species: Mouse
Clone: HMB45
Isotype: IgG1/κ
Species Reactivity: Human. Does not react with dog and rat. Others not known.
Immunogen: Extract of pigmented melanoma metastases from lymph nodes
Molecular Weight: 100kDa
Positive Control: Melanoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Western Blotting
Specifications
Specifications
| Antigen | Melanoma (gp100) Ab-1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | HMB45 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Extract of pigmented melanoma metastases from lymph nodes |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Cancer and Tumor Biology |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.