Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234608MFV450
Description
Melanoma Marker (MART-1 + gp100) Monoclonal antibody specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow CytometrySpecifications
Melanoma Marker (MART-1 + gp100) | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
2315 | |
Store at 4C in the dark. | |
IgG2b κ, IgG1 κ |
Flow Cytometry | |
DT101+ BC199 + HMB45 | |
Flow Cytometry | |
MART1, MART-1 | |
Recombinant Melanoma Marker (MART-1 + gp100) protein (DT101 & BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction