Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody

Catalog No. MS939S Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS939S 1 mL Concentrated
MS939R7 7 mL Prediluted
MS939S0 100 μL Concentrated
MS939S1 500 μL Concentrated
MS939R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS939S Supplier Epredia™ Supplier No. MS939S

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ Melanoma Marker, Pan Ab-1, Mouse Monoclonal Antibody.

Melanoma associated antigens recognized by autologous cytotoxic T cells include MART-1, MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1.

Host Species: Mouse

Clone: HMB45 (gp100), M2-7C10 (MART-1) and M2-9E3 (MART-1)

Isotype: IgG1/κ + IgG2b/κ + IgG2b/κ

Species Reactivity: Human and Horse. Others not known.

Immunogen: Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3)

Molecular Weight: 100kDa (gp100) and 18kDa (MART-1)

Positive Control: Melanoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Melanoma Marker, Pan Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HMB45 (gp100, M2-7C10) MART-1 and M2-9E3 (MART-1)
Conjugate Unconjugated
Host Species Mouse
Immunogen Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant hMART-1 protein (M2-7C10 and M2-9E3)
Purification Method Melanoma
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Primary or Secondary Primary
Target Species Equine, Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.