Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Melanophilin Monoclonal Antibody (OTI6E3)
GREENER_CHOICE

Catalog No. PIMA526992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526992 Supplier Invitrogen™ Supplier No. MA526992
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanophilin Monoclonal Antibody for IHC (P), Western Blot

Slac2-a (for synaptotagmin-like protein (Slp) homolog lacking C2 domains-a) links Rab27A on melanosomes with Myosin VA in melanocytes. Slac2-a binds Myosin VA through a C-terminal region and GTP-bound Rab27A through its synaptotagmin-like protein homology domain (SHD), which is located near the N-terminus. The transport of pigment and cytotoxic granules in melanocytes requires the stable formation of this complex, and thus mutations in the binding domains of the three protein components may cause albinism and/or severe immune disorders.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melanophilin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mlph
Gene Accession No. Q9BV36
Gene Alias 2210418F23Rik; 5031433I09Rik; AW228792; D1Wsu84e; Exophilin-3; l(1)-3Rk; l1Rk3; leaden; leaden protein; Ln; Melanophilin; MGC2771; MGC59733; Mlph; OTTMUSP00000025451; SLAC2A; Slac-2a; SlaC2-a; Slp homolog lacking C2 domains a; synaptotagmin-like protein 2a
Gene Symbols Mlph
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 295-600 of MLPH produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79083
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.