Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Melanopsin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10074460
Description
Melanopsin Polyclonal specifically detects Melanopsin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated.Specifications
Melanopsin | |
Polyclonal | |
Unconjugated | |
PBS, 1mg/ml BSA with 0.05% Sodium Azide | |
OPN4 | |
Synthetic peptide corresponding to residues E(455) Q K S K T P K T K R H L P S L D R R M(474) of rat melanopsin. | |
100 μg | |
Vision | |
94233 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoassay | |
1 mg/mL | |
Western Blot 0.1 - 0.2 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Frozen 1:1000, Knockout Validated | |
melanopsin, MGC142118, MOP, opsin 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human reactivity reported in scientific literature (PMID: 20971788). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction