Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melatonin, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. J6245214PM
Click to view available options
Quantity:
25 g

Melatonine is a hormone usually produced in the pineal gland and released into the blood.

General Description

  • Melatonine is a hormone produced by the pineal gland that shows several biological activities, including the somnolence, sleep-wake cycle, circadian rhythm, stress response, aging, and immunity activities
  • It shows antioxidant, autocoid, paracoid, hormonal, anti- inflammatory and anti-apoptotic activities

Applications

  • Melatonin shows neurohormonal activity and can upregulate antioxidant enzymes and inhibit rat NOS1
  • Melatonin has been reported to be an activator of MEL-1A-R and MEL-1B-R
  • The production and secretion of melatonine are dependent on beta-adrenergic receptor function
  • During melatonine synthesis, the essential amino acid L-tryptophan is used as a precursor
  • This compound inhibits apoptosis in immune cells and neurons, and enhances apoptotic cell death of cancer cells
  • It can inhibit proliferation and metastasis of breast cancer cells by modulating Rho-associated kinase protein-1 expression
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-31-4
Molecular Formula C13H16N2O2
Molecular Weight (g/mol) 232.28 g/mol
MDL Number MFCD00005655
InChI Key DRLFMBDRBRZALE-UHFFFAOYSA-N
Synonym melatonin, melatonine, n-acetyl-5-methoxytryptamine, circadin, 5-methoxy-n-acetyltryptamine, n-2-5-methoxy-1h-indol-3-yl ethyl acetamide, melatol, melatonex, melovine
PubChem CID 896
ChEBI CHEBI:16796
IUPAC Name N-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
SMILES CC(=O)NCCC1=CNC2=C1C=C(C=C2)OC

Specifications

Melting Point 116°C to 120°C
Color White
Quantity 25 g
Beilstein 205542
Merck Index 14,5816
Solubility Information Soluble in ethanol
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Melatonin
EINECSNumber 200-797-7
RTECSNumber AC5955000
TSCA No
Recommended Storage Keep cold

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.