Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Melengestrol Acetate Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175255
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175255 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175255 Supplier Invitrogen™ Supplier No. PA175255
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Working Concentration: 2.5 μg/mL. Sensitivity: 10 ng/mL Melengestrol Acetate produces 94% inhibition in a competitive ELISA employing Melengestrol Acetate polyclonal antibody.

Melengestrol Acetate is a progestogen-like compound used as a cattle feed additive for its growth promoting effects and suppression of estrus (heat) in heifers. Critics have claimed that hormones used as growth promotants in cattle may cause breast cancer. [National Center for Biotechnology Information. PubChem Compound Database; CID=250948].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Melengestrol Acetate
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.6
Gene Alias C25H32O4
Host Species Sheep
Immunogen Melengestrol(3)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.