Learn More
Promega MELK Kinase Enzyme System, 10microg
Catalog No. PRV4150 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4150 10 μg
1 options
Catalog No. PRV4150 Supplier Promega Supplier No. V4150
Recombinant human MELK (1-340) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MELK or maternal embryonic leucine zipper kinase is a member of the Snf1/AMPK kinase family.
  • Easily Screen and Profile MELK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C