Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MELK Rabbit anti-Human, Clone: S06-4D6, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | MELK |
---|---|
Clone | S06-4D6 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
MELK Monoclonal antibody specifically detects MELK in Human samples. It is validated for Western Blot, ELISASpecifications
MELK | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Protein Kinase, Stem Cell Markers | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
9833 | |
IgG | |
Affinity-purified |
S06-4D6 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.11, EC 2.7.11.1, hMELK, HPK38, KIAA0175pEg3 kinase, maternal embryonic leucine zipper kinase, Protein kinase PK38 | |
A synthetic peptide of human MELK (Uniprot#: Q14680) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title